Product Details

SNP ID
rs17883592
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:34442931 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGACTCTACTATATTATCCAAAGC[A/G]TACATTTCATATTTGGTGAAAATCT
Phenotype
MIM: 115500
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAT PubMed Links

Gene Details

Gene
CAT
Gene Name
catalase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001752.3 Intron NP_001743.1

View Full Product Details