Product Details

SNP ID
rs34239319
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:43881131 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGCTTAGTCTGCACCTCAACCGT[G/T]CGGAAAGTGACTGCCCTGTTTACTG
Phenotype
MIM: 610603
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
ALKBH3 PubMed Links

Gene Details

Gene
ALKBH3
Gene Name
alkB homolog 3, alpha-ketoglutaratedependent dioxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139178.3 325 UTR 5 NP_631917.1

View Full Product Details