Product Details

SNP ID
rs34070018
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:43881286 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTATTTTACAGCACTGGTTCTCA[A/G]GCTTTATGTGCATTAAGAATTCCTG
Phenotype
MIM: 610603
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALKBH3 PubMed Links

Gene Details

Gene
ALKBH3
Gene Name
alkB homolog 3, alpha-ketoglutaratedependent dioxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139178.3 Intron NP_631917.1

View Full Product Details