Product Details
- SNP ID
-
hCV59864427
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:27710934 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCACTGCCGTCTACTCGGCCACT[C/G]CGGTCCCGACACCTAGCCTGCGTGA
- Phenotype
-
MIM: 611995
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MRPS35
PubMed Links
Gene Details
- Gene
- MRPS35
- Gene Name
- mitochondrial ribosomal protein S35
There are no transcripts associated with this gene.
View Full Product Details