Product Details

SNP ID
rs35617837
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:14613205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGCTGGCTACCCGTCTGGGTTTT[C/T]GGCTTCTTATCCCTGCTGCCTGTCT
Phenotype
MIM: 601330
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C12orf60 PubMed Links

Gene Details

Gene
C12orf60
Gene Name
chromosome 12 open reading frame 60
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175874.3 3271 Intron NP_787070.2
XM_005253322.4 3271 Intron XP_005253379.1
XM_011520568.2 3271 Intron XP_011518870.1
XM_011520569.2 3271 Intron XP_011518871.1
XM_017018871.1 3271 Intron XP_016874360.1
XM_017018872.1 3271 Intron XP_016874361.1
XM_017018873.1 3271 Intron XP_016874362.1
XM_017018874.1 3271 Intron XP_016874363.1
Gene
GUCY2C
Gene Name
guanylate cyclase 2C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004963.3 3271 Missense Mutation CAA,CGA Q1045R NP_004954.2
XM_011520631.2 3271 Missense Mutation CAA,CGA Q963R XP_011518933.1
Gene
PLBD1-AS1
Gene Name
PLBD1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details