Product Details

SNP ID
rs28990677
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:2878829 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTGTCAAGATTTTCAGCTGCATAA[C/T]TGAAGGAATAATGACACTATTCCCT
Phenotype
MIM: 602341 MIM: 614085
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FOXM1 PubMed Links

Gene Details

Gene
FOXM1
Gene Name
forkhead box M1
There are no transcripts associated with this gene.

Gene
RHNO1
Gene Name
RAD9-HUS1-RAD1 interacting nuclear orphan 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252499.2 Intron NP_001239428.1
NM_001252500.2 Intron NP_001239429.1
NM_001257097.1 Intron NP_001244026.1
NM_001257098.1 Intron NP_001244027.1

View Full Product Details