Product Details

SNP ID
hCV59987394
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:118136242 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACCTCAGCAAGTGGTCCGGGCC[C/G]TTGAGCCTGCAAGAAGTGGACGAGC
Phenotype
MIM: 604591
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
PEBP1 PubMed Links

Gene Details

Gene
PEBP1
Gene Name
phosphatidylethanolamine binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002567.3 359 Silent Mutation CCC,CCG P11P NP_002558.1

View Full Product Details