Product Details

SNP ID
rs11554099
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:118144629 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGGTTTACGAGCAGGACAGGCC[A/G]CTAAAGTGTGACGAGCCCATCCTCA
Phenotype
MIM: 604591 MIM: 616711
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PEBP1 PubMed Links

Gene Details

Gene
PEBP1
Gene Name
phosphatidylethanolamine binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002567.3 716 Silent Mutation CCA,CCG P130P NP_002558.1
Gene
TAOK3
Gene Name
TAO kinase 3
There are no transcripts associated with this gene.

View Full Product Details