Product Details

SNP ID
rs28362515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:110149388 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGACGGCCTTTAAGGTTGTTGCTT[G/T]TGAAGTCAAGTCATTCAGTTTGTGA
Phenotype
MIM: 120130
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
COL4A1 PubMed Links

Gene Details

Gene
COL4A1
Gene Name
collagen type IV alpha 1 chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303110.1 14047 Intron NP_001290039.1
NM_001845.5 14047 UTR 3 NP_001836.3
XM_011521048.2 14047 UTR 3 XP_011519350.1

View Full Product Details