Product Details

SNP ID
rs28595053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40164015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAGTCTTCTATCTAACCATTGCA[A/G]GGCTCTGAATTGCAGAATACGTAAT
Phenotype
MIM: 602860
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BUB1B PubMed Links

Gene Details

Gene
BUB1B
Gene Name
BUB1 mitotic checkpoint serine/threonine kinase B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001211.5 Intron NP_001202.4

View Full Product Details