Product Details

SNP ID
rs34434843
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40241049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATATTTAGGGAGCACTGCTGGGT[C/T]CCAGGCTCCAAGTCACACCCTTTGG
Phenotype
MIM: 608110
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BUB1B-PAK6 PubMed Links

Gene Details

Gene
BUB1B-PAK6
Gene Name
BUB1B-PAK6 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128628.2 Intron NP_001122100.1
NM_001128629.2 Intron NP_001122101.1
Gene
C15orf56
Gene Name
chromosome 15 open reading frame 56
There are no transcripts associated with this gene.

Gene
PAK6
Gene Name
p21 (RAC1) activated kinase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276717.1 Intron NP_001263646.1
NM_001276718.1 Intron NP_001263647.1
NM_020168.5 Intron NP_064553.1

View Full Product Details