Product Details

SNP ID
rs28653441
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43385064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGGGCAAGTAGTGACGATAGGG[G/T]TGTATACTGCTGGTATGCGGGGAAG
Phenotype
MIM: 609610
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TUBGCP4 PubMed Links

Gene Details

Gene
TUBGCP4
Gene Name
tubulin gamma complex associated protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286414.2 Intron NP_001273343.1
NM_014444.4 Intron NP_055259.2
XM_011521454.2 Intron XP_011519756.1
XM_011521455.2 Intron XP_011519757.1
XM_017022078.1 Intron XP_016877567.1

View Full Product Details