Product Details

SNP ID
rs28433760
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43696643 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGAATAACTGAGATGGCACGTCA[A/G]TGCCTGGGATGTGTGCAGTGGGATG
Phenotype
MIM: 613415
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CKMT1A PubMed Links

Gene Details

Gene
CKMT1A
Gene Name
creatine kinase, mitochondrial 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015001.2 Intron NP_001015001.1
NM_001321926.1 Intron NP_001308855.1
NM_001321927.1 Intron NP_001308856.1
NM_001321928.1 Intron NP_001308857.1
NM_001321929.1 Intron NP_001308858.1
XM_005254498.3 Intron XP_005254555.1
XM_017022369.1 Intron XP_016877858.1
XM_017022370.1 Intron XP_016877859.1
XM_017022371.1 Intron XP_016877860.1
Gene
STRCP1
Gene Name
stereocilin pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details