Product Details

SNP ID
rs28616830
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43756558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCAAGAATTCCTTGATCCCCTTA[C/G]GATTTGACCAATCCTAGTATTTTGA
Phenotype
MIM: 602046
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PDIA3 PubMed Links

Gene Details

Gene
PDIA3
Gene Name
protein disulfide isomerase family A member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005313.4 Intron NP_005304.3

View Full Product Details