Product Details

SNP ID
rs28381094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:48331144 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCGGTTCCGGCGCTTAGGGCGCC[A/G]CTAAACTCAGAGCCCGGGAGTCATG
Phenotype
MIM: 601266
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUT PubMed Links

Gene Details

Gene
DUT
Gene Name
deoxyuridine triphosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025248.1 45 Intron NP_001020419.1
NM_001025249.1 45 Intron NP_001020420.1
NM_001948.3 45 Intron NP_001939.1
XM_011521315.2 45 UTR 5 XP_011519617.1
XM_017021988.1 45 Intron XP_016877477.1

View Full Product Details