Product Details

SNP ID
rs28381104
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:48332285 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTGAAGAGACACCCGCCATTTCA[C/T]CCAGTAAGCGGGCCCGGCCTGCGGA
Phenotype
MIM: 601266
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
DUT PubMed Links

Gene Details

Gene
DUT
Gene Name
deoxyuridine triphosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025248.1 376 Missense Mutation CCC,TCC P100S NP_001020419.1
NM_001025249.1 376 UTR 5 NP_001020420.1
NM_001948.3 376 Missense Mutation CCC,TCC P12S NP_001939.1
XM_011521315.2 376 Missense Mutation CCC,TCC P15S XP_011519617.1
XM_017021988.1 376 Missense Mutation CCC,TCC P100S XP_016877477.1

View Full Product Details