Product Details
- SNP ID
-
rs8041008
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:48669897 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGATATACTGTAGTACAATATAGT[A/G]TATATATATATATATATATATATAT
- Phenotype
-
MIM: 613529
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CEP152
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs576225037] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CEP152
- Gene Name
- centrosomal protein 152
View Full Product Details