Product Details

SNP ID
rs28440118
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:50563397 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAAGCAGTCTCAAGAGAGGAACAT[C/T]ATCAGATTTGTTTCAAGAACTATTC
Phenotype
MIM: 605692
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRPM7 PubMed Links

Gene Details

Gene
TRPM7
Gene Name
transient receptor potential cation channel subfamily M member 7
There are no transcripts associated with this gene.

View Full Product Details