Product Details

SNP ID
rs28485857
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:51343332 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAATCTGGACCTAAGCAGAAGT[C/T]TCAAAAAAGGACTGCTATAAGATGA
Phenotype
MIM: 107910 MIM: 608603
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP19A1 PubMed Links

Gene Details

Gene
CYP19A1
Gene Name
cytochrome P450 family 19 subfamily A member 1
There are no transcripts associated with this gene.

Gene
GLDN
Gene Name
gliomedin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181789.2 Intron NP_861454.2
XM_011521501.2 Intron XP_011519803.1
XM_017022121.1 Intron XP_016877610.1
XM_017022122.1 Intron XP_016877611.1
XM_017022123.1 Intron XP_016877612.1
XM_017022124.1 Intron XP_016877613.1
XM_017022125.1 Intron XP_016877614.1
XM_017022126.1 Intron XP_016877615.1

View Full Product Details