Product Details

SNP ID
rs28653485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:51939819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTAAAAGAACTAGCATGGTAATT[C/G]TAAAAGATGGTACAGCCTGTCCTAA
Phenotype
MIM: 610507
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LEO1 PubMed Links

Gene Details

Gene
LEO1
Gene Name
LEO1 homolog, Paf1/RNA polymerase II complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286430.1 Intron NP_001273359.1
NM_001323903.1 Intron NP_001310832.1
NM_001323904.1 Intron NP_001310833.1
NM_138792.3 Intron NP_620147.1
XM_017021911.1 Intron XP_016877400.1

View Full Product Details