Product Details

SNP ID
rs28383497
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:59106691 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTGGGAGCTAAAACTATATTAC[C/T]CAAGAAAACTACTTCCATTCTAAAT
Phenotype
MIM: 602755 MIM: 605840
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCNB2 PubMed Links

Gene Details

Gene
CCNB2
Gene Name
cyclin B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004701.3 Intron NP_004692.1
Gene
RNF111
Gene Name
ring finger protein 111
There are no transcripts associated with this gene.

View Full Product Details