Product Details

SNP ID
rs28718579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:65665727 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGAACTCATCTTGCTCCTCTTG[C/G]TCATGATTGAAAGTCACAGAAAGAT
Phenotype
MIM: 600382 MIM: 603617
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DENND4A PubMed Links

Gene Details

Gene
DENND4A
Gene Name
DENN domain containing 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144823.2 Intron NP_001138295.1
NM_001320835.1 Intron NP_001307764.1
NM_005848.4 Intron NP_005839.3
XM_005254120.4 Intron XP_005254177.1
XM_005254121.3 Intron XP_005254178.1
XM_005254122.4 Intron XP_005254179.1
XM_006720371.3 Intron XP_006720434.1
XM_011521156.2 Intron XP_011519458.1
XM_011521157.2 Intron XP_011519459.1
XM_017021862.1 Intron XP_016877351.1
XM_017021863.1 Intron XP_016877352.1
XM_017021864.1 Intron XP_016877353.1
XM_017021865.1 Intron XP_016877354.1
Gene
SLC24A1
Gene Name
solute carrier family 24 member 1
There are no transcripts associated with this gene.

View Full Product Details