Product Details

SNP ID
rs28414919
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:66296214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTAACACACTGCGAGCCTTTTCAA[A/C]TTCTGTTTTCTGAAATATTTTTAAT
Phenotype
MIM: 614183
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DIS3L PubMed Links

Gene Details

Gene
DIS3L
Gene Name
DIS3 like exosome 3'-5' exoribonuclease
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143688.2 Intron NP_001137160.1
NM_001323936.1 Intron NP_001310865.1
NM_001323937.1 Intron NP_001310866.1
NM_001323938.1 Intron NP_001310867.1
NM_001323939.1 Intron NP_001310868.1
NM_001323940.1 Intron NP_001310869.1
NM_001323941.1 Intron NP_001310870.1
NM_001323943.1 Intron NP_001310872.1
NM_001323944.1 Intron NP_001310873.1
NM_001323945.1 Intron NP_001310874.1
NM_001323946.1 Intron NP_001310875.1
NM_001323948.1 Intron NP_001310877.1
NM_133375.4 Intron NP_588616.1
XM_005254146.3 Intron XP_005254203.1

View Full Product Details