Product Details
- SNP ID
-
rs11542893
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:72241912 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCCGACACTTACACAAAGAAGAA[C/T]CTTGTGCAGACATGGTCGGACACAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PARP6
PubMed Links
Gene Details
- Gene
- PARP6
- Gene Name
- poly(ADP-ribose) polymerase family member 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001323515.1 |
2300 |
Silent Mutation |
AGA,AGG |
R574R |
NP_001310444.1 |
NM_001323516.1 |
2300 |
Silent Mutation |
AGA,AGG |
R574R |
NP_001310445.1 |
NM_001323519.1 |
2300 |
Silent Mutation |
AGA,AGG |
R573R |
NP_001310448.1 |
NM_001323521.1 |
2300 |
Silent Mutation |
AGA,AGG |
R573R |
NP_001310450.1 |
NM_001323522.1 |
2300 |
Silent Mutation |
AGA,AGG |
R593R |
NP_001310451.1 |
NM_001323523.1 |
2300 |
Silent Mutation |
AGA,AGG |
R573R |
NP_001310452.1 |
NM_001323524.1 |
2300 |
Silent Mutation |
AGA,AGG |
R573R |
NP_001310453.1 |
NM_001323525.1 |
2300 |
Silent Mutation |
AGA,AGG |
R594R |
NP_001310454.1 |
NM_001323526.1 |
2300 |
Silent Mutation |
AGA,AGG |
R573R |
NP_001310455.1 |
NM_001323528.1 |
2300 |
Silent Mutation |
AGA,AGG |
R594R |
NP_001310457.1 |
NM_001323530.1 |
2300 |
Silent Mutation |
AGA,AGG |
R574R |
NP_001310459.1 |
NM_001323531.1 |
2300 |
Silent Mutation |
AGA,AGG |
R594R |
NP_001310460.1 |
NM_001323532.1 |
2300 |
Silent Mutation |
AGA,AGG |
R593R |
NP_001310461.1 |
NM_020214.3 |
2300 |
Silent Mutation |
AGA,AGG |
R593R |
NP_064599.2 |
XM_011521805.1 |
2300 |
Silent Mutation |
AGA,AGG |
R459R |
XP_011520107.1 |
XM_017022418.1 |
2300 |
Silent Mutation |
AGA,AGG |
R438R |
XP_016877907.1 |
View Full Product Details