Product Details
- SNP ID
-
rs28756143
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:74904241 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAGCACAGACTAGCAAACACTTAG[C/G]TGGGATCTATCAAAAGGGTTCCTTT
- Phenotype
-
MIM: 154550
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM219B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77509195] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM219B
- Gene Name
- family with sequence similarity 219 member B
- Gene
- MPI
- Gene Name
- mannose phosphate isomerase
There are no transcripts associated with this gene.
View Full Product Details