Product Details

SNP ID
rs28627975
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78094802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGTCAGAGCTGGAGGTGTGGGG[G/T]CACTGGGGACTCCTGCAATTGCCTG
Phenotype
MIM: 605564
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CIB2 PubMed Links

Gene Details

Gene
CIB2
Gene Name
calcium and integrin binding family member 2
There are no transcripts associated with this gene.

Gene
SH2D7
Gene Name
SH2 domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101404.1 Intron NP_001094874.1
XM_011521895.2 Intron XP_011520197.1
XM_011521896.2 Intron XP_011520198.1
XM_011521899.2 Intron XP_011520201.1
XM_017022487.1 Intron XP_016877976.1
XM_017022488.1 Intron XP_016877977.1

View Full Product Details