Product Details

SNP ID
rs28543672
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:80059973 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGCCGGCGCAGCAGGTGGTGGGC[C/T]CGCGGGGGCCGCGGGCGAGAGGGCG
Phenotype
MIM: 610183
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZFAND6 PubMed Links

Gene Details

Gene
ZFAND6
Gene Name
zinc finger AN1-type containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242911.1 Intron NP_001229840.1
NM_001242912.1 Intron NP_001229841.1
NM_001242913.1 Intron NP_001229842.1
NM_001242914.1 Intron NP_001229843.1
NM_001242915.1 Intron NP_001229844.1
NM_001242916.1 Intron NP_001229845.1
NM_001242917.1 Intron NP_001229846.1
NM_001242918.1 Intron NP_001229847.1
NM_001242919.1 Intron NP_001229848.1
NM_019006.3 Intron NP_061879.2
XM_011521684.2 Intron XP_011519986.1

View Full Product Details