Product Details

SNP ID
rs28672389
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:80414146 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTGGCAGGACATGTGAGAAACC[C/T]GGGCACAGAGGCCAAACGTGTATCC
Phenotype
MIM: 606036
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARNT2 PubMed Links

Gene Details

Gene
ARNT2
Gene Name
aryl hydrocarbon receptor nuclear translocator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014862.3 Intron NP_055677.3

View Full Product Details