Product Details
- SNP ID
-
rs28403050
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:97965386 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGTTCAAGGCCGTAGCACTCTGGC[C/G]TGGGCAATAGAGCGAGACCCTGTCT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ARRDC4
PubMed Links
Gene Details
- Gene
- ARRDC4
- Gene Name
- arrestin domain containing 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_183376.2 |
|
Intron |
|
|
NP_899232.2 |
View Full Product Details