Product Details

SNP ID
rs28562768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:89088688 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCACGCCGCAGCCTGAGGGGATCC[C/T]GGGGGGCCTGTCATCTGGCGCCGTG
Phenotype
MIM: 612196
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABHD2 PubMed Links

Gene Details

Gene
ABHD2
Gene Name
abhydrolase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007011.7 Intron NP_008942.3
NM_152924.4 Intron NP_690888.1
XM_005254832.1 Intron XP_005254889.1
XM_005254833.1 Intron XP_005254890.1
XM_011521165.1 Intron XP_011519467.1

View Full Product Details