Product Details

SNP ID
rs28397672
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:90085153 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCGAGCCAGCTAGTGAGGAGGCT[G/T]CCCAGATACAGCTGCCCGCCCCTGG
Phenotype
MIM: 147650
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IDH2 PubMed Links

Gene Details

Gene
IDH2
Gene Name
isocitrate dehydrogenase (NADP(+)) 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289910.1 Intron NP_001276839.1
NM_001290114.1 Intron NP_001277043.1
NM_002168.3 Intron NP_002159.2
Gene
ZNF710
Gene Name
zinc finger protein 710
There are no transcripts associated with this gene.

View Full Product Details