Product Details

SNP ID
rs28406799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:21670475 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGGTCTTCCCTGACCAAACTGT[C/G]TATAATAGTACCTGCTCCTTCTTTG
Phenotype
MIM: 607038
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OTOA PubMed Links

Gene Details

Gene
OTOA
Gene Name
otoancorin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161683.1 Intron NP_001155155.1
NM_144672.3 Intron NP_653273.3
NM_170664.2 Intron NP_733764.1
XM_011545747.2 Intron XP_011544049.1
XM_011545748.2 Intron XP_011544050.2
XM_017022950.1 Intron XP_016878439.1
XM_017022951.1 Intron XP_016878440.1

View Full Product Details