Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001006634.2 | 2945 | UTR 3 | NP_001006635.1 | ||
NM_018054.5 | 2945 | UTR 3 | NP_060524.4 | ||
XM_011545873.2 | 2945 | UTR 3 | XP_011544175.1 | ||
XM_011545874.2 | 2945 | UTR 3 | XP_011544176.1 | ||
XM_011545875.2 | 2945 | UTR 3 | XP_011544177.1 | ||
XM_011545876.2 | 2945 | UTR 3 | XP_011544178.1 | ||
XM_011545877.2 | 2945 | UTR 3 | XP_011544179.1 | ||
XM_011545878.2 | 2945 | UTR 3 | XP_011544180.1 | ||
XM_017023390.1 | 2945 | UTR 3 | XP_016878879.1 |