Product Details

SNP ID
rs17884850
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31093081 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAAAACAACAACAAACAAAAAAC[C/T]TGGGGACCTAGGATGTCTTTAAGGG
Phenotype
MIM: 610561 MIM: 608547
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PRSS53 PubMed Links

Gene Details

Gene
PRSS53
Gene Name
protease, serine 53
There are no transcripts associated with this gene.

Gene
VKORC1
Gene Name
vitamin K epoxide reductase complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001311311.1 Intron NP_001298240.1
NM_024006.5 Intron NP_076869.1
NM_206824.2 Intron NP_996560.1
Gene
ZNF646
Gene Name
zinc finger protein 646
There are no transcripts associated with this gene.

View Full Product Details