Product Details

SNP ID
rs28539217
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:48254811 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGAATTCACTTTTCTCCTTTAG[C/G]CATCCTGTACTGAGCAGTGTTTTGG
Phenotype
MIM: 607040
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ABCC11 PubMed Links

Gene Details

Gene
ABCC11
Gene Name
ATP binding cassette subfamily C member 11
There are no transcripts associated with this gene.

Gene
LONP2
Gene Name
lon peptidase 2, peroxisomal
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300948.1 Intron NP_001287877.1
NM_031490.3 Intron NP_113678.2
XM_017023755.1 Intron XP_016879244.1
XM_017023756.1 Intron XP_016879245.1

View Full Product Details