Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001193522.1 | 583 | Intron | NP_001180451.1 | ||
NM_001193523.1 | 583 | Intron | NP_001180452.1 | ||
NM_001193524.1 | 583 | Intron | NP_001180453.1 | ||
NM_024519.3 | 583 | Intron | NP_078795.2 | ||
XM_011523321.1 | 583 | UTR 5 | XP_011521623.1 | ||
XM_011523322.1 | 583 | Intron | XP_011521624.1 | ||
XM_011523324.2 | 583 | Intron | XP_011521626.1 | ||
XM_011523325.1 | 583 | Intron | XP_011521627.1 | ||
XM_011523326.1 | 583 | Intron | XP_011521628.1 | ||
XM_017023663.1 | 583 | Intron | XP_016879152.1 | ||
XM_017023664.1 | 583 | Intron | XP_016879153.1 |