Product Details

SNP ID
rs28455856
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:69586201 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCATTTATGGTTTTCTTCCTTC[C/T]GTTTTCAGAGAGTGTGAATTTCTGA
Phenotype
MIM: 604708
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NFAT5 PubMed Links

Gene Details

Gene
NFAT5
Gene Name
nuclear factor of activated T-cells 5, tonicity-responsive
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113178.2 Intron NP_001106649.1
NM_006599.3 Intron NP_006590.1
NM_138713.3 Intron NP_619727.2
NM_138714.3 Intron NP_619728.2
NM_173214.2 Intron NP_775321.1
NM_173215.2 Intron NP_775322.1
XM_006721125.3 Intron XP_006721188.1
XM_011522817.2 Intron XP_011521119.1
XM_011522818.2 Intron XP_011521120.1
XM_011522819.2 Intron XP_011521121.1
XM_011522820.2 Intron XP_011521122.1
XM_011522822.2 Intron XP_011521124.1
XM_017022870.1 Intron XP_016878359.1
XM_017022871.1 Intron XP_016878360.1
XM_017022872.1 Intron XP_016878361.1
XM_017022873.1 Intron XP_016878362.1

View Full Product Details