Product Details

SNP ID
rs28461645
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:81449772 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTGGAGGTGGTGGAGGGGAGGGG[C/T]TGAGACAGAGAGGAGAAAAAGTCAA
Phenotype
MIM: 610112
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CMIP PubMed Links

Gene Details

Gene
CMIP
Gene Name
c-Maf inducing protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030629.2 Intron NP_085132.1
NM_198390.2 Intron NP_938204.2
XM_005256179.4 Intron XP_005256236.4
XM_005256181.2 Intron XP_005256238.1
XM_005256182.1 Intron XP_005256239.1
XM_011523352.1 Intron XP_011521654.1
XM_011523353.1 Intron XP_011521655.1
XM_017023733.1 Intron XP_016879222.1

View Full Product Details