Product Details

SNP ID
rs28486087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89929319 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAAAGAATAAAACTTAGCAATAA[C/G]AAAAATGCACAAAAGATGGAAAGAA
Phenotype
MIM: 155555 MIM: 602661
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MC1R PubMed Links

Gene Details

Gene
MC1R
Gene Name
melanocortin 1 receptor
There are no transcripts associated with this gene.

Gene
TUBB3
Gene Name
tubulin beta 3 class III
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197181.1 Intron NP_001184110.1
NM_006086.3 Intron NP_006077.2

View Full Product Details