Product Details

SNP ID
rs28370891
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82718913 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGTATTTTTCTGACAGGCCAGAA[A/G]AATGTTTGAAGGTGAGATGGCAAGT
Phenotype
MIM: 611683
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FN3KRP PubMed Links

Gene Details

Gene
FN3KRP
Gene Name
fructosamine 3 kinase related protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024619.3 199 Missense Mutation AAA,AGA K50R NP_078895.2
Gene
LOC101929552
Gene Name
uncharacterized LOC101929552
There are no transcripts associated with this gene.

View Full Product Details