Product Details

SNP ID
rs28634753
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:16683794 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATATGCAAGCACATGAAGCCGAG[C/T]GTGAAGCCGAGCGTGAAGCCGAGCG
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CCDC144A PubMed Links

Gene Details

Gene
CCDC144A
Gene Name
coiled-coil domain containing 144A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014695.2 Intron NP_055510.1
XM_011524089.2 Intron XP_011522391.1
XM_017025429.1 Intron XP_016880918.1
XM_017025430.1 Intron XP_016880919.1

View Full Product Details