Product Details
- SNP ID
-
rs3209163
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:6800754 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACTGGCTGCTAGCCTACCTTGCGA[G/C]ATTGTGGGTGATCTCTTGAACCTCC
- Phenotype
-
MIM: 609106
MIM: 609002
- Polymorphism
- G/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FBXO39
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17804647] are located under a probe and SNP(s) [rs2271233] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FBXO39
- Gene Name
- F-box protein 39
There are no transcripts associated with this gene.
- Gene
- TEKT1
- Gene Name
- tektin 1
View Full Product Details