Product Details

SNP ID
rs4417599
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8040662 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAAAGAAAGAAAGAAAAGAAAGA[A/G]AAAGAAACTGCTTTAAATGCCTTAC
Phenotype
MIM: 603697
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALOX15B PubMed Links

Gene Details

Gene
ALOX15B
Gene Name
arachidonate 15-lipoxygenase, type B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039130.1 Intron NP_001034219.1
NM_001039131.1 Intron NP_001034220.1
NM_001141.2 Intron NP_001132.2

View Full Product Details