Product Details

SNP ID
rs28372844
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40287981 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGAGAGGCTGCGTGTGAGAGACGT[A/G]AGAAGGATCCTGCACTGAGGAGGTG
Phenotype
MIM: 602627 MIM: 609692
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC6 PubMed Links

Gene Details

Gene
CDC6
Gene Name
cell division cycle 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001254.3 293 UTR 5 NP_001245.1
XM_011525541.1 293 UTR 5 XP_011523843.1
XM_011525542.1 293 UTR 5 XP_011523844.1
Gene
WIPF2
Gene Name
WAS/WASL interacting protein family member 2
There are no transcripts associated with this gene.

View Full Product Details