Product Details
- SNP ID
-
rs28630617
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:40749391 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCATCAATTCTAGGATCACCATATG[A/G]TTTTCTTTAGCTCTTTGGTAGTTTC
- Phenotype
-
MIM: 616646
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT25
PubMed Links
Gene Details
- Gene
- KRT25
- Gene Name
- keratin 25
View Full Product Details