Product Details

SNP ID
rs35468867
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42017602 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGTCCACGGTGGTCTCCGCGCCG[G/T]GGCGGGAGCCCAGGGGCTGTGCCGG
Phenotype
MIM: 601964 MIM: 604497
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC7 PubMed Links

Gene Details

Gene
DNAJC7
Gene Name
DnaJ heat shock protein family (Hsp40) member C7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144766.2 27 Intron NP_001138238.1
NM_003315.3 27 UTR 5 NP_003306.3
XM_011525167.2 27 Intron XP_011523469.1
XM_011525168.2 27 Intron XP_011523470.1
XM_011525169.2 27 Intron XP_011523471.1
XM_017024994.1 27 Intron XP_016880483.1
Gene
NKIRAS2
Gene Name
NFKB inhibitor interacting Ras like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001349.2 27 UTR 5 NP_001001349.1
NM_001144927.1 27 Intron NP_001138399.1
NM_001144928.1 27 Intron NP_001138400.1
NM_001144929.1 27 Intron NP_001138401.1
NM_017595.5 27 Intron NP_060065.2
XM_005257251.1 27 Intron XP_005257308.1
Gene
ZNF385C
Gene Name
zinc finger protein 385C
There are no transcripts associated with this gene.

View Full Product Details