Product Details

SNP ID
rs1141073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42022415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATACCAGGTTCGGAGATGATCGA[G/T]ACGCAGGAGGACATCTACGTGGGCT
Phenotype
MIM: 601964 MIM: 604497
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC7 PubMed Links

Gene Details

Gene
DNAJC7
Gene Name
DnaJ heat shock protein family (Hsp40) member C7
There are no transcripts associated with this gene.

Gene
NKIRAS2
Gene Name
NFKB inhibitor interacting Ras like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001349.2 176 Missense Mutation GAG,GAT E37D NP_001001349.1
NM_001144927.1 176 Missense Mutation GAG,GAT E37D NP_001138399.1
NM_001144928.1 176 Missense Mutation GAG,GAT E37D NP_001138400.1
NM_001144929.1 176 Missense Mutation GAG,GAT E37D NP_001138401.1
NM_017595.5 176 Missense Mutation GAG,GAT E37D NP_060065.2
XM_005257251.1 176 Missense Mutation GAG,GAT E37D XP_005257308.1
Gene
ZNF385C
Gene Name
zinc finger protein 385C
There are no transcripts associated with this gene.

View Full Product Details