Product Details

SNP ID
rs34108277
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:45070071 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCATTCCGGTTTTAAGGAGTTT[A/C]GTATACCAGAGCTTTTGTGACCTCT
Phenotype
MIM: 160993
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DCAKD PubMed Links

Gene Details

Gene
DCAKD
Gene Name
dephospho-CoA kinase domain containing
There are no transcripts associated with this gene.

Gene
NMT1
Gene Name
N-myristoyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021079.4 Intron NP_066565.1
XM_011524858.2 Intron XP_011523160.1

View Full Product Details