Product Details
- SNP ID
-
rs17850101
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:45149518 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGACTTCCCGCCGCCGGCAGAAGTG[A/G]AACCGACGCCCGAGGCCGAGCTGCT
- Phenotype
-
MIM: 607328
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ACBD4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12051846] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ACBD4
- Gene Name
- acyl-CoA binding domain containing 4
There are no transcripts associated with this gene.
- Gene
- HEXIM1
- Gene Name
- hexamethylene bisacetamide inducible 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006460.2 |
2202 |
Missense Mutation |
AAA,GAA |
K110E |
NP_006451.1 |
View Full Product Details