Product Details

SNP ID
rs28533679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:64553050 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAAACGTAATACATACTTTTTAC[A/G]TATATAACAAAAAAATCAAACATTA
Phenotype
MIM: 605532
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMURF2 PubMed Links

Gene Details

Gene
SMURF2
Gene Name
SMAD specific E3 ubiquitin protein ligase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022739.3 Intron NP_073576.1
XM_005257585.3 Intron XP_005257642.1
XM_011525128.1 Intron XP_011523430.1

View Full Product Details